2-Isopropoxyphenol ≥97%

AC20922-0250
200034-852EA 107 USD
200034-852
2-Isopropoxyphenol ≥97%
2-Isopropoxyphenol
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 63…66 °C (0.15 mmHg)
Melting Pt: 206…207 °C
Density: 1.03 g/cm³ (20 °C)
MDL Number: MFCD00002184
CAS Number: 4812-20-8
EINECS: 225-379-1

Order Now

Specification Test Results

Appearance Clear light yellow to red-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5140 to 1.5160 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR