Cyclobutylamine 98%

Danger

Synonyms: CyclobutanamineAminocyclobutaneCyclobutylamine

AC20911-0010 AC20911-0050
102776-112EA 187.38 USD
102776-112 200018-604
Cyclobutylamine 98%
Cyclobutylamine
Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 81.4 °C (1013 hPa)
Density: 0.83 g/cm³
Flash Pt: –4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001328
CAS Number: 2516-34-9
EINECS: 219-736-0
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4360 to 1.4400 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR