1-Chloro-4-fluorobenzene 98%

Warning

Synonyms: Chloro-1-fluoro-4-benzène4-Chlorofluorobenzene

AC20820-0050 AC20820-0250
200045-936EA 19.4 USD
200045-936 200027-850
1-Chloro-4-fluorobenzene 98%
1-Chloro-4-fluorobenzene
Formula: C₆H₄ClF
MW: 130.55 g/mol
Boiling Pt: 129 °C (1013 hPa)
Melting Pt: –21.5 °C
Density: 1.226 g/cm³
Flash Pt: 29 °C
MDL Number: MFCD00000603
CAS Number: 352-33-0
EINECS: 206-521-1
UN: 1993

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.4960 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR