2-(Cyclohexylamino)ethanesulfonic acid (CHES) 98+% for biochemistry

Synonyms: N-Cyclohexyltaurine

AC20818-0250 AC20818-5000 AC20818-1000
200005-996EA 100 USD
200005-996 103511-298 103511-296
2-(Cyclohexylamino)ethanesulfonic acid (CHES) 98+% for biochemistry
2-(Cyclohexylamino)ethanesulphonic acid (CHES)
Formula: C₈H₁₇NO₃S
MW: 207.29 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003835
CAS Number: 103-47-9
EINECS: 203-115-6
Merck Index: 13,02063

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
UV (0.1 M in water)
UV at 260 nm A: ≤0.05
UV at 280 nm A: ≤0.04
pH 5 to 6 (1 % in water)
Titration with NaOH ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR