Spironolactone 99%

Danger

AC20746-0010
200007-734EA 58.5 USD
200007-734
Spironolactone 99%
Spironolactone
Formula: C₂₄H₃₂O₄S
Melting Pt: 198…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00082250
CAS Number: 52-01-7
EINECS: 200-133-6
Merck Index: 12,08917

Order Now

Specification Test Results

Appearance White to yellowish-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105 °C, 2 h)
HPLC ≥98.5 %
Specific optical rotation -41° to -45° (20 °C, 589 nm) (c=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR