4-Fluorocinnamic acid 99%

Warning

AC20711-0250 AC20711-0050
200032-492EA 34.3 USD
200032-492 200038-464
4-Fluorocinnamic acid 99%
4-Fluorocinnamic acid
Formula: C₉H₇FO₂
MW: 166.15 g/mol
Melting Pt: 207…210 °C
MDL Number: MFCD00004395
CAS Number: 459-32-5
EINECS: 207-288-9

Order Now

Specification Test Results

Appearance White to off-white Crystalline solid
Infrared spectrum Conforms
Melting point 207 to 210 °C
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR