Copper(II) chloride dihydrate 98%, pure

Danger

Synonyms: Cupric chloride dihydrateCopper dichloride dihydrate

AC20634-0025
103526-612EA 228 USD
103526-612
Copper(II) chloride dihydrate 98%, pure
Copper (II) chloride dihydrate
Formula: CuCl₂·2H₂O
MW: 170.48 g/mol
Melting Pt: 100 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 13,02660

Order Now

Specification Test Results

Appearance Blue-green to blue Crystalline powder or needles
Titration Complexometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR