Aluminium ammonium sulfate dodecahydrate 99%, pure

Warning

Synonyms: Aluminium ammonium bis(sulphate) dodecahydrateAluminium ammonium bis(sulfate) dodecahydrate

AC20626-0100 AC20626-0025
103463-918EA 1050.8 USD
103463-918 200048-512
Aluminium ammonium sulfate dodecahydrate 99%, pure
Aluminium ammonium sulphate dodecahydrate
Formula: AlNH₄(SO₄)₂·12H₂O
MW: 453.33 g/mol
Boiling Pt: 200 °C (760 mmHg)
Melting Pt: 93.5 °C
Density: 1.64 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00051042
CAS Number: 7784-26-1
EINECS: 232-055-3
Merck Index: 13,00325

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Complexometric 98.5 to 102.5 %
Loss on drying 45 % to 48 % (2 g, 300 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR