1-Heptanesulfonic acid sodium salt 98%

Warning

Synonyms: Sodium 1-heptanesulfonateSodium 1-heptanesulphonateSodium heptane-1-sulfonateSodium heptane-1-sulphonate

AC20613-0250 AC20613-5000 AC20613-1000
103516-736EA 1234 USD
103516-736 200002-384 200008-674
1-Heptanesulfonic acid sodium salt 98%
1-Heptanesulphonic acid sodium salt
Formula: H₃C(CH₂)₅CH₂SO₃Na
MW: 202.25 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007543
CAS Number: 22767-50-6
EINECS: 245-210-5
https://us.vwr.com/stibo/web/std.lang.all/07/91/2300791.jpg

Order Now

Specification Test Results

Appearance White Powder or flakes
Infrared spectrum Conforms
Loss on drying ≤9 % (100 °C, 3 h) (vacuum)
Solubility (10 % in water) Clear colorless
Titration after Ion exchange ≥97.5 % (on dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR