Kojic acid 99%

Warning

Synonyms: 5-Hydroxy-2-hydroxymethyl-4H-pyran-4-one5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one5-Hydroxy-2-hydroxymethyl-γ-pyrone5-Hydroxy-2-hydroxymethyl-4-pyrone

AC20522-5000 AC20522-0050 AC20522-0250 AC20522-1000
200025-612EA 91.5 USD
200025-612 103509-696 200019-422 200012-914
Kojic acid 99%
Kojic acid
Formula: C₆H₆O₄
MW: 142.11 g/mol
Melting Pt: 151…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00006580
CAS Number: 501-30-4
EINECS: 207-922-4
Merck Index: 13,05334

Order Now

Specification Test Results

Appearance White to beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 151 to 156 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR