3-Hydroxy-p-anisic acid 99%

Warning

Synonyms: Isovanillic acid3-Hydroxy-4-methoxybenzoic acid

AC20503-0050 AC20503-0250
200009-956EA 84.4 USD
200009-956 200063-488
3-Hydroxy-p-anisic acid 99%
3-Hydroxy-p-anisic acid
Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 327 °C (1013 hPa)
Melting Pt: 245…250 °C
MDL Number: MFCD00002507
CAS Number: 645-08-9
EINECS: 211-430-5

Order Now

Specification Test Results

Appearance Beige to grey-brown Powder
Infrared spectrum Conforms
Melting point 245 to 250 °C
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR