3-(Indol-3-yl)propionic acid 99%

Warning

Synonyms: 3-Indolepropionic acidIndole-3-propionic acid

AC20499-0100 AC20499-0010 AC20499-0500
103509-690EA 318 USD
103509-690 103509-688 200009-980
3-(Indol-3-yl)propionic acid 99%
3-(Indol-3-yl)propionic acid
Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
Melting Pt: 130…136 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005660
CAS Number: 830-96-6
EINECS: 212-600-1

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 130 to 136 °C
Water ≤0.5 % (K.F.)
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR