1-Fluoro-3-iodobenzene ≥99%

Supplier: ACROS ORGANICS MS

Warning

Synonyms: 3-Fluoroiodobenzene

AC20448-0050 AC20448-0250
200015-796EA 29.75 USD
200015-796 200021-834
1-Fluoro-3-iodobenzene ≥99%
1-Fluoro-3-iodobenzene
Formula: C₆H₄FI
MW: 222 g/mol
Boiling Pt: 183 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.89 g/cm³ (20 °C)
MDL Number: MFCD00001044
CAS Number: 1121-86-4
EINECS: 214-339-9

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5830 to 1.5860 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer Copper

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR