Tetraphenylphosphonium bromide 99%

Warning

AC20402-0250 AC20402-0050 AC20402-1000
103516-710EA 179 USD
103516-710 200020-910 200020-908
Tetraphenylphosphonium bromide 99%
Tetraphenylphosphonium bromide
Formula: C₂₄H₂₀BrP
MW: 419.3 g/mol
Melting Pt: 295…299 °C
Flash Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00011915
CAS Number: 2751-90-8
EINECS: 220-393-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 296 to 300 °C
Titration Argentometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR