1-Aminocyclohexanecarboxylic acid 98%

Warning

Synonyms: 1-Amino-1-cyclohexanecarboxylic acid1-Aminocyclohexane-1-carboxylic acid

AC20395-0250 AC20395-1000
200033-866EA 212.5 USD
200033-866 200033-864
1-Aminocyclohexanecarboxylic acid 98%
1-Aminocyclohexanecarboxylic acid
Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00001487
CAS Number: 2756-85-6
EINECS: 220-411-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR