Barium chloride dihydrate 99+% for analysis
|
Danger
|
AC20313-0025
103524-130EA
181
USD
103524-130
Barium chloride dihydrate 99+% for analysis
Barium chloride dihydrate
Formula:
BaCl₂·2H₂O MW: 244.27 g/mol Boiling Pt: 1560 °C (760 mmHg) Melting Pt: 962 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149154 CAS Number: 10326-27-9 EINECS: 233-788-1 UN: 1564 ADR: 6.1,III Merck Index: 13,00974 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Titration Argentometric | ≥99.0 % |
Loss on drying | 14.0 % to 16.0 % (150 °C) |
pH | 5.2 to 8.2 (5 % at 25 °C) |
Calcium (Ca) | ≤500 ppm |
Iron (Fe) | ≤2 ppm |
Potassium (K) | ≤25 ppm |
Sodium (Na) | ≤50 ppm |
Strontium (Sr) | ≤1000 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...