Trichloroacetamide 98+%

Warning

Synonyms: 2,2,2-Trichloroacetamide

AC20292-1000 AC20292-0250
200028-058EA 23.4 USD
200028-058 200028-060
Trichloroacetamide 98+%
Trichloroacetamide
Formula: C₂H₂Cl₃NO
MW: 162.4 g/mol
Boiling Pt: 238…240 °C (760 mmHg)
Melting Pt: 139…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00008009
CAS Number: 594-65-0
EINECS: 209-849-3

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 139 to 143 °C
HPLC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR