6-Aminonicotinic acid 98%

Warning

Synonyms: 6-Aminopyridine-3-carboxylic acid6-Amino-3-pyridinecarboxylic acid

AC20254-0250 AC20254-0050 AC20254-0010
103465-700EA 29.3 USD
103465-700 200048-284 200024-150
6-Aminonicotinic acid 98%
6-Aminonicotinic acid
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006326
CAS Number: 3167-49-5
EINECS: 221-630-4
Merck Index: 13,00455

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR