2,4-Toluenediisocyanate 75%, Technical Grade

Danger

Synonyms: 2,4-TDI4-Methyl-m-phenylene diisocyanateTolylene 2,4-diisocyanate

AC20250-0025 AC20250-2500 AC20250-0010
200027-006EA 138 USD
200027-006 200020-970 103531-774
2,4-Toluenediisocyanate 75%, Technical Grade
2,4-Toluenediisocyanate
Formula: C₉H₆N₂O₂
Boiling Pt: 115…120 °C (10 mmHg)
Melting Pt: 12.5…13.5 °C
Density: 1.22 g/cm³
Flash Pt: 127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002011
CAS Number: 584-84-9
EINECS: 247-722-4
UN: 2078
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5660 to 1.5700 (20 °C, 589 nm)
NMR 70 to 90 Tolylene 2,4-diisocyanate
NMR 10 to 30 % Tolylene 2,6-diisocyanate

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR