Tannic acid 95%

Synonyms: TanninGallotannin

AC20242-0050 AC20242-0010 AC20242-0051 AC20242-5000
200014-626EA 157 USD
200014-626 200014-624 200020-786 103509-648
Tannic acid 95%
Tannic acid
Formula: C₇₆H₅₂O₄₆
MW: 1701.2 g/mol
Melting Pt: 218 °C (dec.)
Flash Pt: 198 °C
Storage Temperature: Ambient
MDL Number: MFCD00066397
CAS Number: 1401-55-4
EINECS: 215-753-2
Merck Index: 12,09221

Order Now

Specification Test Results

Appearance Yellow to brown Powder
Infrared spectrum Conforms
Assay ≥94 % (on dry substance)
Loss on drying ≤12.0 %
Sulfated ash ≤1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR