Antimony(III) chloride 99.5%

Danger

Synonyms: Antimony trichloride

AC20168-0100 AC20168-5000
102777-862EA 1038.8 USD
102777-862 200011-256
Antimony(III) chloride 99.5%
Antimony (III) chloride
Formula: SbCl₃
MW: 228.12 g/mol
Boiling Pt: 223.5 °C (1013 hPa)
Melting Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00011212
CAS Number: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8,II
Merck Index: 13,00713

Order Now

Specification Test Results

Appearance Colorless to white to off-white Adhering crystals and/or chunks (May have a pink hue)
Titration Iodimetric ≥99.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR