Nickel 99.97% (trace metals basis), spheres 5 --> 11 mm

Danger

200020-070EA 94.23 USD
200020-070
Nickel 99.97% (trace metals basis), spheres 5 --> 11 mm
Nickel
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1455 °C
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
ADR: 4.1,III
Merck Index: 12,06582

Order Now

Specification Test Results

Appearance Spheres
Assay ≥99.92 % (Ni)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR