Antimony(V) chloride ≥99%

Supplier: ACROS ORGANICS MS

Danger

Synonyms: Antimony pentachloride

AC20121-0010 AC20121-0500 AC20121-2500
200011-258EA 405.91 USD
200011-258 200005-776 200018-182
Antimony(V) chloride ≥99%
Antimony (V) chloride
Formula: Cl₅Sb
MW: 299.02 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 3 °C
Density: 2.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011213
CAS Number: 7647-18-9
EINECS: 231-601-8
UN: 1730
ADR: 8,II
Merck Index: 13,00701

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid (May develop some turbidity or precipitate)
Assay ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR