Potassium cyanate 97%, pure

Warning

AC20111-1000
103516-670EA 33.6 USD
103516-670
Potassium cyanate 97%, pure
Potassium cyanate
Formula: CKNO
MW: 81.12 g/mol
Melting Pt: 315 °C
Storage Temperature: Ambient
MDL Number: MFCD00011411
CAS Number: 590-28-3
EINECS: 209-676-3
Merck Index: 12,07788

Order Now

Specification Test Results

Appearance White Crystalline powder and/or chunks
Titration Argentometric ≥96.0 %
Potassium carbonate ≤3 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR