4-Fluoro-3-nitrobenzotrifluoride 96%

Warning

Synonyms: 4-Fluoro-3-Nitrobenzotrifluoride

AC20061-1000 AC20061-0050
102774-852EA 105 USD
102774-852 200029-816
4-Fluoro-3-nitrobenzotrifluoride 96%
4-Fluoro-3-nitrobenzotrifluoride
Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
Boiling Pt: 92 °C (15 mmHg)
Density: 1.49 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00007059
CAS Number: 367-86-2
EINECS: 206-702-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4630 (20 °C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR