Sulfur 99.999% (trace metals basis)

Warning

AC19993-0100 AC19993-0500 AC19993-2500
200038-838EA 328 USD
200038-838 200014-610 200014-608
Sulfur 99.999% (trace metals basis)
Sulphur
Formula: S
MW: 32.07 g/mol
Boiling Pt: 445 °C (1013 hPa)
Melting Pt: 114 °C
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00085316
CAS Number: 7704-34-9
EINECS: 231-722-6
UN: 1350
ADR: 4.1,III
Merck Index: 12,09142

Order Now

Specification Test Results

Appearance Yellow Pieces
Particle size ≤12 mm
Total trace metal impurities ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR