Selenious acid 98%

Danger

Synonyms: Selenous acid

AC19887-0010
103526-530EA 673 USD
103526-530
Selenious acid 98%
Selenious acid
Formula: H₂O₃Se
MW: 128.97 g/mol
Melting Pt: 70 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011331
CAS Number: 7783-00-8
EINECS: 231-974-7
UN: 3283
ADR: 6.1,II
Merck Index: 14,08430

Order Now

Specification Test Results

Appearance White to off-white to light pink Crystalline powder or crystals and/or chunks
Titration Iodometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR