Sodium periodate 99% for analysis
|
Danger
|
Synonyms:
Sodium metaperiodate
AC19838-1000
AC19838-5000
200002-208EA
384
USD
200002-208
200002-206
Sodium periodate 99% for analysis
Sodium periodate
Formula:
NaIO₄ MW: 213.89 g/mol Melting Pt: 300 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003534 CAS Number: 7790-28-5 EINECS: 232-197-6 UN: 3085 ADR: 5.1,I Merck Index: 12,08786 |
Specification Test Results
Appearance | White to off-white Crystals |
Titration Iodometric | ≥98.5 % |
pH | 3.4 to 4.5 (5 % at 20 °C) |
Chloride (Cl) | ≤300 ppm |
Iodide (I) | ≤10 ppm |
Nitrate (NO3) | ≤2000 ppm |
Sulfate (SO4) | ≤200 ppm |
Iron (Fe) | ≤10 ppm |
Lead (Pb) | ≤20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...