α-Bromo-4-fluorotoulene 97%

Danger

Synonyms: α-Bromo-p-fluorotoulenep-Fluorobenzyl bromide4-Fluorobenzyl bromide

AC19797-0050 AC19797-0250
200017-532EA 39.1 USD
200017-532 200005-434
α-Bromo-4-fluorotoulene 97%
α-Bromo-4-fluorotoulene
Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 85 °C (15 mmHg)
Density: 1.517 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000359
CAS Number: 459-46-1
EINECS: 207-291-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5464 to 1.5484 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR