α-Bromo-2-fluorotoulene 98%

Danger

AC19796-0050 AC19796-0250
200005-048EA 118 USD
200005-048 103530-054
α-Bromo-2-fluorotoulene 98%
α-Bromo-2-fluorotoulene
Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 84…85 °C (15 mmHg)
Density: 1.567 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000324
CAS Number: 446-48-0
EINECS: 207-169-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5495 to 1.5515 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR