4-Fluorobenzamide 98%

Warning

AC19790-0050 AC19790-0250
200023-784EA 38.5 USD
200023-784 200047-892
4-Fluorobenzamide 98%
4-Fluorobenzamide
Formula: C₇H₆FNO
MW: 139.13 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 154…158 °C
MDL Number: MFCD00007992
CAS Number: 824-75-9
EINECS: 212-535-9

Order Now

Specification Test Results

Appearance White to beige to light brown Powder
Infrared spectrum Conforms
Melting point 154 to 158 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR