3-Methoxybenzyl chloride 97%

Danger

Synonyms: 1-(Chloromethyl)-3-methoxybenzene3-(Chloromethyl)anisole

AC19786-1000 AC19786-0250 AC19786-0050
103530-052EA 250 USD
103530-052 200019-812 200025-930
3-Methoxybenzyl chloride 97%
3-Methoxybenzyl chloride
Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 124 °C (13 mmHg)
Density: 1.07 g/cm³
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000907
CAS Number: 824-98-6
EINECS: 212-541-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5430 to 1.5450 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR