Germanium(IV) chloride 99.99% (trace metals basis)

Danger

Synonyms: Germanium tetrachlorideTetrachlorogermane

AC19715-1000
200031-460EA 678 USD
200031-460
Germanium(IV) chloride 99.99% (trace metals basis)
Germanium (IV) chloride
Formula: Cl₄Ge
MW: 214.42 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –49.5 °C
Density: 1.84 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011029
CAS Number: 10038-98-9
EINECS: 233-116-7
UN: 2922
ADR: 8,I
Merck Index: 12,04418

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Total trace metal impurities ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR