Cadmium(II) chloride hemipentahydrate 99+%

Danger

AC19702-2500
200042-808EA 128 USD
200042-808
Cadmium(II) chloride hemipentahydrate 99+%
Cadmium (II) chloride hemipentahydrate
Formula: CdCl₂·2.5H₂O
MW: 228.35 g/mol
Melting Pt: 34 °C
MDL Number: MFCD00799071
CAS Number: 7790-78-5
EINECS: 233-296-7
UN: 2570
ADR: 6.1,III
Merck Index: 13,01617

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Titration Complexometric 79.5 to 81.0 % (CdCl2)
Nitrate and Nitrite (as NO3) ≤0.003 %
Sulfate (SO4) ≤50 ppm
Ammonium (NH4) ≤50 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤50 ppm
Zinc (Zn) ≤500 ppm
Insoluble matter ≤0.005 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR