Aluminium acetylacetonate 97%

Danger

Synonyms: Aluminium tris(2,4-pentanedionato-O,O')Aluminum acetylacetonateAluminum 2,4-pentanedionateTris(2,4-pentanedionato-O,O')aluminiumAluminium 2,4-pentanedionate

AC19697-0050 AC19697-5000 AC19697-1000
200005-698EA 31.7 USD
200005-698 102775-502 200030-382
Aluminium acetylacetonate 97%
Aluminium acetylacetonate
Formula: C₁₅H₂₁AlO₆
MW: 324.31 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 190…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00000013
CAS Number: 13963-57-0
EINECS: 237-741-6
UN: 3467
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 190 to 195 °C
Titration Complexometric ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR