Iodine monochloride ≥98%

Supplier: ACROS ORGANICS MS

Danger

Synonyms: ChloroiodideIodine chlorideIodochloride

AC19696-5000 AC19696-1000
200006-702EA 447.79 USD
200006-702 103529-106
Iodine monochloride ≥98%
Iodine monochloride
Formula: ClI
MW: 162.36 g/mol
Boiling Pt: 97°C (760mmHg)
Melting Pt: 26 °C
Density: 3.1 g/cm³ (29 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00011354
CAS Number: 7790-99-0
EINECS: 232-236-7
UN: 1792
ADR: 8,II
Merck Index: 13,05039

Order Now

Specification Test Results

Appearance Dark red-brown to brown to black Liquid or solid
Titration Iodometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR