D-(+)-Raffinose pentahydrate 99+%

Synonyms: Raffinose, 5-HydrateGossypose pentahydrateMelitose pentahydrate(3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-((((3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol pentahydrateD-(+)-Raffinose pentahydrateRaffinose, PentahydrateRaffinose pentahydrateMelitriose pentahydrate

AC19567-0250 AC19567-1000
200002-102EA 252 USD
200002-102 200002-100
D-(+)-Raffinose pentahydrate 99+%
D(+)-Raffinose pentahydrate
Formula: C₁₈H₃₂O₁₆·5H₂O
MW: 594.52 g/mol
Melting Pt: 78…80.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00071590
CAS Number: 17629-30-0
EINECS: 208-146-9
Merck Index: 12,08279

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water 14 to 16 % (K.F.)
Sulfated ash ≤0.1 % (on dry basis)
Solubility (10 % in water) Clear colorless
HPLC ≥99.0 % (ELSD)
Specific optical rotation +103.0° to +107.0° (20 °C, 589 nm) (c=10, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR