Nickel 99.9% (trace metals basis), powder -325 mesh

Danger

AC19361-1000 AC19361-0250
200007-150EA 48.25 USD
200007-150 200020-068
Nickel 99.9% (trace metals basis), powder -325 mesh
Nickel
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 2371 °C (1013 hPa)
Melting Pt: 1455 °C
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Appearance Grey Powder
Particle size ≤44 µm
Total trace metal impurities ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR