Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manganese(IV) oxide ≥99.99% (trace metals basis)

Supplier: ACROS ORGANICS MS

Warning
Synonyms: Manganese dioxideManganese(IV) oxide, activatedMangan(IV)-oxidManganese peroxide

AC19347-0050 AC19347-1000 AC19347-0250
200025-744EA 68.89 USD InStock
200025-744 103509-416 102776-344
Manganese(IV) oxide ≥99.99% (trace metals basis)
Manganese (IV) oxide


Formula: MnO₂
MW: 86.94 g/mol
Melting Pt: 535 °C (dec.)
Density: 5.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6
Merck Index: 13,05753
Order Now
Specification Test Results
Appearance Grey to black Powder or crystals and/or chunks
Total trace metal impurities ≤100 ppm


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty