Polyethylene glycol 6,000

Synonyms: PEG 6000PEG 6,000

AC19228-0010 AC19228-0051
200002-068EA 67 USD
200002-068 103511-186
Polyethylene glycol 6,000
Polyethylene glycol 6,000
Formula: H(OCH₂CH₂)nOH
MW: 6000 g/mol
Melting Pt: 55…60 °C
Storage Temperature: Ambient
MDL Number: MFCD01779614
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Specification Test Results

Appearance White Powder or flakes
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Hydroxyl value 17 to 20 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR