Polyethylene glycol 600

Synonyms: PEG 600

AC19224-0051
103511-180EA 398.46 USD
103511-180
Polyethylene glycol 600
Polyethylene glycol 600
Formula: H(OCH₂CH₂)nOH
MW: 600 g/mol
Melting Pt: 17…22 °C
Density: 1.1265 g/cm³
Flash Pt: 252 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Specification Test Results

Appearance Clear colourless to almost colourless Viscous liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.4660 to 1.4700 (20 °C, 589 nm)
Water ≤2.0 % (K.F.)
Hydroxyl value 178 to 197 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR