Chlorophenol red, pure indicator grade

Warning

Synonyms: 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

AC19004-0500
103516-570EA 277 USD
103516-570
Chlorophenol red, pure indicator grade
Chlorophenol red
Formula: C₁₉H₁₂Cl₂O₅S
MW: 423.27 g/mol
Melting Pt: 261 °C
Storage Temperature: Ambient
MDL Number: MFCD00005877
CAS Number: 4430-20-0
EINECS: 224-619-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR