ω-Bromo-2,4-dimethoxyacetophenone 98%

Danger

AC19001-0010
200016-362EA 89.9 USD
200016-362
ω-Bromo-2,4-dimethoxyacetophenone 98%
ω-Bromo-2,4-dimethoxyacetophenone
Formula: C₁₀H₁₁BrO₃
MW: 259.1 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 101…105 °C
MDL Number: MFCD00000197
CAS Number: 60965-26-6
EINECS: 262-542-6
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR