3-Fluoropyridine 99%

Danger

AC18830-0050 AC18830-0010
200047-478EA 37 USD
200047-478 102775-024
3-Fluoropyridine 99%
3-Fluoropyridine
Formula: C₅H₄FN
MW: 97.09 g/mol
Boiling Pt: 107 °C (1013 hPa)
Density: 1.13 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00006374
CAS Number: 372-47-4
EINECS: 206-755-4
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR