5-Methylbenzotriazole

Synonyms: 5-Methyl-1H-benzotriazole

AC18800-0100
200036-272EA 34.9 USD
200036-272
5-Methylbenzotriazole
5-Methylbenzotriazole
Formula: C₇H₇N₃
MW: 133.15 g/mol
Boiling Pt: 361 °C (1013 hPa)
Melting Pt: 85 °C
MDL Number: MFCD00005702
CAS Number: 136-85-6
EINECS: 205-265-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR