4-Phenylcyclohexanone 98+%

Synonyms: 1-Oxo-4-phenylcyclohexane4-Phenylcyclohexan-1-one

AC18794-0100 AC18794-0500
103510-718EA 46.4 USD
103510-718 200017-692
4-Phenylcyclohexanone 98+%
4-Phenylcyclohexanone
Formula: C₁₂H₁₄O
MW: 174.24 g/mol
Melting Pt: 77…80 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00001641
CAS Number: 4894-75-1
EINECS: 225-517-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR