4-Isopropylbenzyl alcohol 97%

Warning

Synonyms: (4-Isopropylphenyl)methanol

AC18780-0250 AC18780-5000 AC18780-1000
200047-942EA 76 USD
200047-942 200042-070 103510-716
4-Isopropylbenzyl alcohol 97%
4-Isopropylbenzyl alcohol
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 248 °C (760 mmHg)
Melting Pt: 28 °C
Density: 0.98 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00004663
CAS Number: 536-60-7
EINECS: 208-640-4
Merck Index: 13,02646

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR