cis-1,4-Dichloro-2-butene 95%

Danger

AC18760-5000 AC18760-1000 AC18760-0250
200011-686EA 119 USD
200011-686 200030-734 103531-718
cis-1,4-Dichloro-2-butene 95%
cis-1,4-Dichloro-2-butene
Formula: C₄H₆Cl₂
MW: 125 g/mol
Boiling Pt: 152 °C (758 mmHg)
Melting Pt: –48 °C
Density: 1.18 g/cm³
Flash Pt: 55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062950
CAS Number: 1476-11-5
EINECS: 216-021-5
UN: 3489
ADR: 6.1,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR