2-Nitroimidazole 98%

Danger

Synonyms: 2-Nitro-1H-imidazole

AC18757-2500
200022-878EA 102 USD
200022-878
2-Nitroimidazole 98%
2-Nitroimidazole
Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
Melting Pt: 287 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005185
CAS Number: 527-73-1
EINECS: 208-425-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,00923

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR