3,4-Dimethoxyphenol 97%

Warning

AC18670-0100 AC18670-0500
200029-088EA 165 USD
200029-088 103509-404
3,4-Dimethoxyphenol 97%
3,4-Dimethoxyphenol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Melting Pt: 77…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00008390
CAS Number: 2033-89-8
EINECS: 217-995-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR